Span® 40

Supplier: TCI America
Warning

Synonyms: Sorbitan monopalmitate, Arlacel 40

S0061-500G
TCS0061-500GEA 60.38 USD
TCS0061-500G
Span® 40
Span® 40

CAS Number: 26266-57-9
MDL Number: MFCD00080946
Form: Pellet
Color: Pale Yellow
Melting point (°C): 47
Flash Point (°C): 113


Other Product Names / Synonyms: Arlacel 40, Sorbitan Monopalmitate

Formula: C₂₂H₄₂O₆
Boiling Pt: ∼100 °C (1013 hPa)
Melting Pt: 47 °C
Density: 1 g/cm³ (20 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00080946
CAS Number: 26266-57-9
EINECS: 247-568-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR