Sorbic acid ≥99.0%
Supplier: TCI America
|
Warning
|
Synonyms:
(E,E)-2,4-Hexadienoic acid, (2E,4E)-2,4-Hexadienoic acid, trans,trans-2,4-Hexadienoic acid, 2,4-Hexadienoic acid, (2E,4E)-Hexa-2,4-dienoic acid
S0053-500G
S0053-25G
TCS0053-500GEA
66.14
USD
TCS0053-500G
TCS0053-25G
Sorbic acid ≥99.0%
Sorbic acid
CAS Number: 110-44-1
MDL Number: MFCD00002703
Molecular Formula: C6H8O2
Molecular Weight: 112.13
Purity/Analysis Method:
Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: 2,4-Hexadienoic Acid
Formula:
CH₃CH=CHCH=CHCOOH MW: 112.13 g/mol Boiling Pt: 228 Melting Pt: 134 °C Density: 1.204 g/cm³ (20 °C) Flash Pt: 127 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002703 CAS Number: 110-44-1 EINECS: 203-768-7 Merck Index: 13,08793 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...