β-Sitosterol 40,0% (contains Campesterol)

Supplier: TCI America

Synonyms: α-Dihydrofucosterol, β-Sitosterol, 5-Stigmasten-3β-ol, 22,23-Dihydrostigmasterol, 24α-Ethylcholesterol

S0040-25G
TCS0040-025GEA 75.28 USD
TCS0040-025G
β-Sitosterol 40,0% (contains Campesterol)
beta-Sitosterol

CAS Number: 83-46-5 MDL Number: MFCD00003631 Molecular Formula: C29H50O Molecular Weight: 414.72 Purity/Analysis Method: 40.0% (GC) Form: Crystal Color: White Melting point (°C): 140 Specific rotation [a]20/D: -35 deg (C=2, Dioxane)

Formula: C₂₉H₅₀O
MW: 414.72 g/mol
Melting Pt: 140 °C
MDL Number: MFCD00003631
CAS Number: 83-46-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=460b99d68008b870011f386fbac8219854edc481&vid=460b99d68008b870011f386fbac8219854edc481&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 40.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR