rPSL1a-LecBeads [for Siaα(2-6)Gal]
Supplier: TCI America
The rPSL1a is an Escherichia coli recombinant of carbohydrate-binding protein (Lectin) derived from Polyporus squamosus. This lectin preferably binds to a [Siaα(2-6)Gal] glycan epitope (It very slightly interacts with lactose.).
The LecBeads is a lectin-immobilized agarose which is capable of capturing glycoproteins via binding to its specific epitope(s), and it is prepared by a covalent coupling method between lectin and agarose.
The Lectin-catch method using LecBeads (as shown in Application tab) is available for selective separation of glycoconjugates (glycoproteins, glycolipids, and so on). This rPSL1a-LecBeads (1 ml gel) shows a binding capacity of about 10 mg or more human Transferrin [hTF] through its binding to a terminal [Siaα(2-6)Gal] glycan epitope on hTF.
- Preparation : Recombinant PSL1a is covalently coupled with agarose gel
- Quantity : 1 ml gel/2 ml (1:1, v/v slurry) suspended in PBS, pH7.5
- Protein concentration : about 10 mg/ml gel
- Binding capacity : >10 mg of glycoprotein (human Transferrin) /ml gel
- Preservative : 0.02% [w/v] Sodium azide
- Stabilizer : None
- Store at 2-10 °C. (Avoid freezing).
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...