[RuCl(p-cymene)((R)-binap)]Cl ≥90.0%
Supplier: TCI America
Synonyms:
Chloro[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl](p-cymene)ruthenium(II) chloride [RuCl(p-cymene)((R)-binap)]Cl, (R)-RuCl[(p-cymene)(BINAP)]Cl, Chloro[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl](p-cymene)ruthenium(II) Chloride, [(R)-Binap RuCl p-cymene]Cl
CAS Number: 145926-28-9
MDL Number: MFCD00134456
Molecular Formula: C54H46Cl2P2Ru
Molecular Weight: 928.88
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Asymmetric Synthesis
Other Product Names / Synonyms: Chloro[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl](p-cymene)ruthenium(II) Chloride
Formula:
C₅₄H₄₆Cl₂P₂Ru |
MDL Number:
MFCD00134456 CAS Number: 145926-28-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...