Rosiglitazone ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
R0106-1G
R0106-200MG
TCR0106-1GEA
219.7
USD
TCR0106-1G
TCR0106-200MG
Rosiglitazone ≥98.0% (by HPLC, titration analysis)
Rosiglitazone
CAS Number: 122320-73-4
MDL Number: MFCD00871760
Molecular Formula: C18H19N3O3S
Molecular Weight: 357.43
Purity/Analysis Method:
Formula:
C₁₈H₁₉N₃O₃S Melting Pt: 156 °C |
MDL Number:
MFCD00871760 CAS Number: 122320-73-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...