Rhodium 5% on carbon wetted with ca. 55% water

Supplier: TCI America
Danger

R0075-1G
TCR0075-1GEA 58 USD
TCR0075-1G
Rhodium 5% on carbon wetted with ca. 55% water
Rhodium

CAS Number: 7440-16-6 Molecular Formula: Rh Molecular Weight: 102.91 Form: Crystals Color: Black


Product Categories: Synthetic Organic Chemistry, Functional Group Transformation and Introducing Reaction, Metal Catalysts for Organic Synthesis

Formula: Rh
MW: 102.9 g/mol
MDL Number: MFCD00011201
CAS Number: 7440-16-6
UN: 1378
ADR: 4.2,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d954dfb13b47747df6f991f3e930b42d93645a02&vid=0cb1d796477672e1d6b52ccad6f1de98c644333c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Black powder to crystal
Content(Rhodium) 4.9 to 5.1 %(calcd.on dried substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR