Rose Bengal
Supplier: TCI America
R0041-25G
TCR0041-025GEA
76.03
USD
TCR0041-025G
Rose Bengal
Rose Bengal
CAS Number: 632-69-9
MDL Number: MFCD00151169
Molecular Formula: C20H4Cl4I4O5
Molecular Weight: 1017.63
Form: Crystal
Color: Deep Yellow Red
Other Product Names / Synonyms: Rose Bengal
Formula:
C₂₀H₄Cl₄I₄O₅ |
MDL Number:
MFCD00151169 CAS Number: 632-69-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...