Rhodamine B ≥95.0%

Supplier: TCI America
Danger

Synonyms: 9-(2-Carboxyphenyl)-3,6-bis(diethylamino)xanthylium chloride, [9-(2-Carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium chloride

R0040-25G R0040-250G
TCR0040-025GEA 54.92 USD
TCR0040-025G TCR0040-250G
Rhodamine B ≥95.0%
Rhodamine B

CAS Number: 81-88-9 MDL Number: MFCD00011931 Molecular Formula: C28H31ClN2O3 Molecular Weight: 479.02 Purity/Analysis Method: 95.0% (HPLC) Form: Crystal Color: Yellow Melting point (°C): 211 Lambda max.: 554 nm (H2O)


Product Categories: Materials Chemistry, Xanthene Dyes (Functional Organic Dyes), Bioscience, Stains & Dyes, Fluorescent Stains, Analytical Science, Fluorescent Dyes Other Product Names / Synonyms: Basic Violet 10

Formula: C₂₈H₃₁ClN₂O₃
Melting Pt: 211 °C
Density: 1.31 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011931
CAS Number: 81-88-9
EINECS: 201-383-9
Merck Index: 13,08266
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=daa0764afa6a24891ef507201d36103bd6e3c98d&vid=daa0764afa6a24891ef507201d36103bd6e3c98d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR