Rutin hydrate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
R0035-25G
TCR0035-025GEA
67.41
USD
TCR0035-025G
Rutin hydrate ≥98.0% (by titrimetric analysis)
Rutin hydrate
CAS Number: 207671-50-9
MDL Number: MFCD00006830
Molecular Formula: C27H30O16
Molecular Weight: 610.52
Purity/Analysis Method:
Formula:
C₂₇H₃₀O₁₆ |
MDL Number:
MFCD00006830 CAS Number: 207671-50-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...