D-(-)-Ribose ≥98.0%

Supplier: TCI America

R0025-250G R0025-25G
TCR0025-250GEA 108.2 USD
TCR0025-250G TCR0025-025G
D-(-)-Ribose ≥98.0%
D(-)-Ribose

CAS Number: 50-69-1 MDL Number: MFCD00135453 Molecular Formula: C5H10O5 Molecular Weight: 150.13 Purity/Analysis Method: 98.0% (GC) Form: Crystal Color: White Specific rotation [a]20/D: -21 deg (C=1, H2O)

Formula: C₅H₁₀O₅
Boiling Pt: 331 °C (1013 hPa)
Melting Pt: 90…95 °C
Density: 0.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00135453
CAS Number: 50-69-1
EINECS: 200-059-4
Merck Index: 12,08371
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47fb4068d14650f4f7e6b4d08c25bc54826877a2&vid=47fb4068d14650f4f7e6b4d08c25bc54826877a2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR