Ribonucleic acid sodium salt (from yeast)

Supplier: TCI America

Synonyms: Sodium Nucleinate, Nucleic Acid Sodium Salt, Sodium Ribonucleinate

R0024-25G
TCR0024-025GEA 139 USD
TCR0024-025G
Ribonucleic acid sodium salt (from yeast)
Ribonucleic acid sodium salt

MDL Number: MFCD00148224
Form: Crystal
Color: Pale Gray


Other Product Names / Synonyms: Nucleic Acid Sodium Salt, Sodium Nucleinate, Sodium Ribonucleinate

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR