Ribonucleic acid (from yeast)

Supplier: TCI America

Synonyms: Nucleic Acid

R0022-25G
TCR0022-025GEA 86.18 USD
TCR0022-025G
Ribonucleic acid (from yeast)
Ribonucleic acid

MDL Number: MFCD00132195
Form: Crystal
Color: White


Other Product Names / Synonyms: Nucleic Acid

MDL Number: MFCD00132195
CAS Number: 63231-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR