Trifluoroacetic acid ≥99.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Q0053-25ML
TCQ0053-025MLEA 39.69 USD
TCQ0053-025ML
Trifluoroacetic acid ≥99.0% (by titrimetric analysis)
Trifluoroacetic acid

[Sequenation Reagent] CAS Number: 76-05-1 MDL Number: MFCD00004169 Molecular Formula: C2HF3O2 Molecular Weight: 114.02 Purity/Analysis Method: 99.0% (T) Form: Clear Liquid Color: Colorless Specific Gravity (20/20): 1.49


Other Product Names / Synonyms: TFA

Formula: C₂HF₃O₂
Boiling Pt: 71.8 °C (1013 hPa)
Melting Pt: –15 °C
Density: 1.490 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004169
CAS Number: 76-05-1
EINECS: 200-929-3
UN: 2699
ADR: 8,I
Merck Index: 13,09754
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=138549c41fa521ad9e755511c7f8483acb5e20a7&vid=fdc0b10b610bc02d6be84bfd961e0775a08265bb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR