Benzene ≥99.5%

Supplier: TCI America
Danger

Synonyms: Cyclohexatriene

Q0038-500ML
TCQ0038-500MLEA 58.2 USD
TCQ0038-500ML
Benzene ≥99.5%
Benzene

[Sequencing Solvent] CAS Number: 71-43-2 MDL Number: MFCD00003009 Molecular Formula: C6H6 Molecular Weight: 78.11 Purity/Analysis Method: 99.5% (GC) Form: Clear Liquid Color: Colorless Boiling point (°C): 80 Flash Point (°C): -10 Freezing point (°C): 5 Specific Gravity (20/20): 0.88

Formula: C₆H₆
MW: 78.11 g/mol
Boiling Pt: 80 °C
Melting Pt: 5.5 °C
Density: 0.88 (20 °C)
Flash Pt: –10 °C
Storage Temperature: Ambient
MDL Number: MFCD00003009
CAS Number: 71-43-2
EINECS: 200-753-7
UN: 1114
ADR: 3,II
REACH: 01-2119447106-44
Merck Index: 13,01066
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44907c7eb2d6e32d5119dde250cf928d00bf6a31&vid=b57af89d29cc988ea5dca51503cb20131b60dc2f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.5% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR