Plerixafor (AMD3100) ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: AMD 3100, Mezobil, JM 3100, Plerixafor, 1,4-Bis((1,4,8,11-tetraazacyclotetradecan-1-yl)methyl)benzene, 1,1'-(1,4-phenylenebis(methylene))bis(1,4,8,11-tetraazacyclotetradecane)octahydrochloride dihydrate, 1,4-Bis[(1,4,8,11-tetraazacyclotetradecan-1-yl)methyl]benzene

P2678-250MG P2678-50MG
TCP2678-250MGEA 298 USD
TCP2678-250MG TCP2678-50MG
Plerixafor (AMD3100) ≥98.0% (by GC)
Plerixafor (AMD3100)
Formula: C₂₈H₅₄N₈
Storage Temperature: Refrigerator
MDL Number: MFCD05662218
CAS Number: 110078-46-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 131.0 to 135.0 deg-C
Water max. 13.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR