(R)-Methyl-2-amino-2-phenylacetate hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: D-(-)-2-Phenylglycine methyll ester hydrochloride, (R)-2-Phenylglycine methyl ester hydrochloride, Methyl (R)-(-)-2-Amino-2-phenylacetate hydrochloride, H-D-Phg-OMe·HCl, H-D-Phg-OMe.HCl, (R)-(-)-2-Amino-2-phenylacetic acid methyll ester hydrochloride

P2592-25G
TCP2592-25GEA 34 USD
TCP2592-25G
(R)-Methyl-2-amino-2-phenylacetate hydrochloride ≥98.0% (by HPLC, titration analysis)
(R)-Methyl-2-amino-2-phenylacetate hydrochloride
Formula: C₉H₁₁NO₂·HCl
MW: 201.7 g/mol
MDL Number: MFCD00137487
CAS Number: 19883-41-1
EINECS: 243-399-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -117 to -129 deg (C=1, water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR