Procarbazine hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Danger

P2550-100MG P2550-500MG
TCP2550-100MGEA 42.25 USD
TCP2550-100MG TCP2550-500MG
Procarbazine hydrochloride ≥98.0% (by HPLC, titration analysis)
Procarbazine hydrochloride
Formula: C₁₂H₁₉N₃O·HCl
Melting Pt: 226°C
MDL Number: MFCD00072082
CAS Number: 366-70-1
EINECS: 206-678-6
Merck Index: 14,07758

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR