Poly(2-ethyl-2-oxazoline) n=∼100, ULTROXA®

Supplier: TCI America

P2507-500MG
TCP2507-500MGEA 166.86 USD
TCP2507-500MG
Poly(2-ethyl-2-oxazoline) n=∼100, ULTROXA®
Poly(2-ethyl-2-oxazoline), M.W. ~200,000
Formula: (C₅H₉NO)x
Storage Temperature: Refrigerator
CAS Number: 25805-17-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Average molecular weight 8,000 to 12,000
Molecular weight distribution max. 1.2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR