4-(1H-Pyrazol-1-yl)benzoic acid ≥98.0% (by GC)

Supplier: TCI America
Warning

P2499-1G P2499-5G
TCP2499-1GEA 58 USD
TCP2499-1G TCP2499-5G
4-(1H-Pyrazol-1-yl)benzoic acid ≥98.0% (by GC)
4-(1H-Pyrazol-1-yl)benzoic acid
Formula: C₁₀H₈N₂O₂
MDL Number: MFCD02682056
CAS Number: 16209-00-0

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR