2-Phenylethylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

P2476-1G P2476-5G
TCP2476-1GEA 29 USD
TCP2476-1G TCP2476-5G
2-Phenylethylboronic acid (contains varying amounts of Anhydride)
2-Phenylethylboronic acid
Formula: C₈H₁₁BO₂
Melting Pt: 88°C
MDL Number: MFCD01631226
CAS Number: 34420-17-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 98.0 to 113.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR