2,2-Dimethyl-N-pyridin-2-ylpropionamide ≥98.0% (by GC)

Supplier: TCI America

P2385-25G P2385-5G
TCP2385-25GEA 92 USD
TCP2385-25G TCP2385-5G
2,2-Dimethyl-N-pyridin-2-ylpropionamide ≥98.0% (by GC)
2,2-Dimethyl-N-pyridin-2-ylpropionamide
Formula: C₁₀H₁₄N₂O
MDL Number: MFCD00460277
CAS Number: 86847-59-8

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 71.0 to 75.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR