Phenyl propargyl ether ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
(Prop-2-ynyloxy)benzene, (Prop-2-yn-1-yloxy)benzene
P2222-25G
P2222-5G
TCP2222-25GEA
367
USD
TCP2222-25G
TCP2222-5G
Phenyl propargyl ether ≥98.0% (by GC)
Phenyl propargyl ether
Formula:
C₉H₈O Boiling Pt: 83°C Flash Pt: 76°C Storage Temperature: Refrigerator |
MDL Number:
MFCD00048107 CAS Number: 13610-02-1 EINECS: 237-095-5 |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...