Palladium(II) acetate ≥98.0% (by titrimetric analysis), purified by sublimation

Supplier: TCI America

Synonyms: Palladium diacetate, Palladium (II) acetate

P2161-1G
TCP2161-1GEA 313.8 USD
TCP2161-1G
Palladium(II) acetate ≥98.0% (by titrimetric analysis), purified by sublimation
Palladium (II) acetate

CAS Number: 3375-31-3
MDL Number: MFCD00012453
Molecular Formula: C4H6O4Pd
Molecular Weight: 224.51
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: Yellow
Lambda max.: 400 nm (EtOH)


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Cross-coupling Reaction using Transition Metal Catalysts, C-H Bond Activation Reaction, Oxidation
Other Product Names / Synonyms: Acetic Acid Palladium(II) Salt

Formula: C₄H₆O₄Pd
MDL Number: MFCD00012453
CAS Number: 3375-31-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=88773b8cc00d12095cacd7c46f30cd2c3eee25c6&vid=8f77c1c73a9045587915840f6e266f43c54d7890&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR