N-(3-Pyridyl)pivalamide ≥98.0% (by GC, titration analysis)

Supplier: TCI America

P2109-25G P2109-5G
TCP2109-25GEA 156.9 USD
TCP2109-25G TCP2109-5G
N-(3-Pyridyl)pivalamide ≥98.0% (by GC, titration analysis)
N-(3-Pyridyl)pivalamide

CAS Number: 70298-88-3 MDL Number: MFCD00996243 Molecular Formula: C10H14N2O Molecular Weight: 178.24 Purity/Analysis Method: 98.0% (GC,T) Form: Crystal Color: White Melting point (°C): 130


Other Product Names / Synonyms: 2,2-Dimethyl-N-(3-pyridyl)propionamide

Formula: C₁₀H₁₄N₂O
Melting Pt: 130 °C
MDL Number: MFCD00996243
CAS Number: 70298-88-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d380b26754c0f831b7595a965fc6068ede001731&vid=d380b26754c0f831b7595a965fc6068ede001731&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR