Pyrene ≥98.0% (by GC), purified by sublimation

Supplier: TCI America

P2072-1G
TCP2072-1GEA 42.9 USD
TCP2072-1G
Pyrene ≥98.0% (by GC), purified by sublimation
Pyrene

CAS Number: 129-00-0
MDL Number: MFCD00004136
Molecular Formula: C16H10
Molecular Weight: 202.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 404
Melting point (°C): 151
Flash Point (°C): 199
Lambda max.: 330 nm (EtOH)


Product Categories: Materials Chemistry, Nanocarbon Unit Structures (Nanocarbon Materials), Highly Purified Reagents
Other Product Names / Synonyms: Benzo[def]phenanthrene

Formula: C₁₆H₁₀
Boiling Pt: 404 °C
Melting Pt: 151 °C
Flash Pt: 199 °C
MDL Number: MFCD00004136
CAS Number: 129-00-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5938a5200052f7cf6bec77bba54b9406967bd68d&vid=bb9c0578054e3dcb94994918b09c8b169ca7ef34&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR