Pantoprazole sulfide ≥98.0%
Supplier: TCI America
Synonyms:
5-Difluoromethoxy-2-[(3,4-dimethoxy-2-pyridyl)methyl]thio-1H-benzimidazole, 5-(Difluoromethoxy)-2-(((3,4-dimethoxypyridin-2-yl)methyl)thio)-1H-benzo[d]imidazole, 5-(Difluoromethoxy)-2-[[(3,4-dimethoxy-2-pyridyl)methyl]thio]benzimidazole
CAS Number: 102625-64-9
MDL Number: MFCD07368273
Molecular Formula: C16H15F2N3O3S
Molecular Weight: 367.37
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Fluorinated Bioactive Compounds, Bioscience, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: 5-(Difluoromethoxy)-2-[[(3,4-dimethoxy-2-pyridyl)methyl]thio]benzimidazole
Formula:
C₁₆H₁₅F₂N₃O₃S MW: 367.37 g/mol Melting Pt: 118 °C |
MDL Number:
MFCD07368273 CAS Number: 102625-64-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...