Platinum(II) chloride ≥95.0% (by gravimetric analysis)

Supplier: TCI America

Synonyms: Platinum dichloride

P1920-1G
TCP1920-1GEA 175 USD
TCP1920-1G
Platinum(II) chloride ≥95.0% (by gravimetric analysis)
Platinum (II) chloride

CAS Number: 10025-65-7 MDL Number: MFCD00011181 Molecular Formula: PtCl2 Molecular Weight: 265.98 Purity/Analysis Method: 95.0% (W) Form: Crystal Color: Pale Yellow Melting point (°C): 581

Formula: Cl₂Pt
MW: 265.99 g/mol
Melting Pt: 581 °C
MDL Number: MFCD00011181
CAS Number: 10025-65-7
UN: 3260
ADR: 8,III

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (W)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR