Pirfenidone ≥98.0%

Supplier: TCI America

Synonyms: 5-Methyl-1-phenylpyridin-2(1H)-one, 5-Methyl-1-phenyl-2-pyridone

P1871-100MG P1871-1G
TCP1871-100MGEA 65.24 USD
TCP1871-100MG TCP1871-1G
Pirfenidone ≥98.0%
Pirfenidone

CAS Number: 53179-13-8 MDL Number: MFCD00866047 Molecular Formula: C12H11NO Molecular Weight: 185.23 Purity/Analysis Method: 98.0% (GC) Form: Crystal Color: White Melting point (°C): 109


Product Categories: Bioscience, Anti-inflammatory Ingredients, Anti-inflammatory Ingredients, Other Pharmaceutical Ingredients Other Product Names / Synonyms: 5-Methyl-1-phenyl-2-pyridone

Formula: C₁₂H₁₁NO
MW: 185.23 g/mol
Melting Pt: 109 °C
MDL Number: MFCD00866047
CAS Number: 53179-13-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a91166278bd695246852b2a0eb8e28d42519fe3&vid=1a91166278bd695246852b2a0eb8e28d42519fe3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR