Pirfenidone ≥98.0%
Supplier: TCI America
Synonyms:
5-Methyl-1-phenylpyridin-2(1H)-one, 5-Methyl-1-phenyl-2-pyridone
P1871-100MG
P1871-1G
TCP1871-100MGEA
65.24
USD
TCP1871-100MG
TCP1871-1G
Pirfenidone ≥98.0%
Pirfenidone
CAS Number: 53179-13-8
MDL Number: MFCD00866047
Molecular Formula: C12H11NO
Molecular Weight: 185.23
Purity/Analysis Method:
Product Categories: Bioscience, Anti-inflammatory Ingredients, Anti-inflammatory Ingredients, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: 5-Methyl-1-phenyl-2-pyridone
Formula:
C₁₂H₁₁NO MW: 185.23 g/mol Melting Pt: 109 °C |
MDL Number:
MFCD00866047 CAS Number: 53179-13-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...