2-Piperazinone ≥98.0%

Supplier: TCI America

Synonyms: Piperazin-2-one

P1867-1G P1867-5G
TCP1867-1GEA 24 USD
TCP1867-1G TCP1867-5G
2-Piperazinone ≥98.0%
2-Piperazinone

CAS Number: 5625-67-2 MDL Number: MFCD01318687 Molecular Formula: C4H8N2O Molecular Weight: 100.12 Purity/Analysis Method: 98.0% (GC) Form: Crystal Color: Slightly Pale Yellow Boiling point (°C): 164 Melting point (°C): 134


Other Product Names / Synonyms: Piperazin-2-one, 2-Oxopiperazine

Formula: C₄H₈N₂O
MW: 100.12 g/mol
Boiling Pt: 164 °C
Melting Pt: 134 °C
MDL Number: MFCD01318687
CAS Number: 5625-67-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=54b877030ecbb069c1421d1083370c3eb7f4507f&vid=54b877030ecbb069c1421d1083370c3eb7f4507f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR