1,10-Phenanthroline monohydrate ≥99.0% (by titrimetric analysis) for biochemical research

Supplier: TCI America

Synonyms: 1,10-Phenanthroline hydrate

P1826-5G
TCP1826-5GEA 47.83 USD
TCP1826-5G
1,10-Phenanthroline monohydrate ≥99.0% (by titrimetric analysis) for biochemical research
1,10-Phenanthroline monohydrate

[for Biochemical Research]
CAS Number: 5144-89-8
MDL Number: MFCD00149973
Molecular Formula: C12H8N2
Molecular Weight: 180.21
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Color: White
Melting point (°C): 102

Formula: C₁₂H₈N₂·1H₂O
MW: 198.22 g/mol
Melting Pt: 102 °C
MDL Number: MFCD00149973
CAS Number: 5144-89-8
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=885a0f0fae4ef9fd25401df9e424ed9b90c01f94&vid=1e4e13c368085e4d5577a8d867ac5ca0920fdeeb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR