1,2,2,3-Propanetetracarbonitrile ≥96.0% (by GC)
Supplier: TCI America
CAS Number: 1274904-48-1
Molecular Formula: C7H4N4
Molecular Weight: 144.14
Purity/Analysis Method:
Product Categories: Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: 1,2,2,3-Tetracyanopropane
Formula:
C₇H₄N₄ |
CAS Number:
1274904-48-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...