Platinium(IV) oxide ≥85.0%

Supplier: TCI America

Synonyms: Platinum dioxide, Adam's type catalyst

P1720-200MG
TCP1720-200MGEA 69 USD
TCP1720-200MG
Platinium(IV) oxide ≥85.0%
Platinium (IV) oxide

CAS Number: 1314-15-4 MDL Number: MFCD00011184 Molecular Formula: O2Pt Molecular Weight: 227.08 Purity/Analysis Method: 85.0% (W) Form: Crystal Color: Gray

Formula: O₂Pt
MW: 227.08
Melting Pt: 450
Density: 11.8
MDL Number: MFCD00011184
CAS Number: 1314-15-4
UN: 1479
ADR: 5.1 II
Merck Index: 12,07686
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2c708cda79b2bab677db3cd2784c7b0be202094&vid=b2c708cda79b2bab677db3cd2784c7b0be202094&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 85.0% (W)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR