2-(4-Pyridyl)benzimidazole ≥98.0%
Supplier: TCI America
Synonyms:
2-(Pyridin-4-yl)-1H-benzo[d]imidazole
P1658-1G
TCP1658-1GEA
146.5
USD
TCP1658-1G
2-(4-Pyridyl)benzimidazole ≥98.0%
2-(4-Pyridyl)benzimidazole
CAS Number: 2208-59-5
Molecular Formula: C12H9N3
Molecular Weight: 195.23
Purity/Analysis Method:
Formula:
C₁₂H₉N₃ Melting Pt: 218 °C |
MDL Number:
MFCD00453948 CAS Number: 2208-59-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...