Perylene ≥99.0%, purified by sublimation

Supplier: TCI America

P1629-1G
TCP1629-1GEA 59 USD
TCP1629-1G
Perylene ≥99.0%, purified by sublimation
Perylene

CAS Number: 198-55-0
MDL Number: MFCD00004142
Molecular Formula: C20H12
Molecular Weight: 252.32
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 278


Product Categories: Materials Chemistry, Organic Light-Emitting Diode (OLED) Materials, Molecular Conductors, Nanocarbon Unit Structures (Nanocarbon Materials), Highly Purified Reagents
Other Product Names / Synonyms: Dibenz[de,kl]anthracene, Peri-dinaphthalene

Formula: C₂₀H₁₂
MW: 252.32 g/mol
Boiling Pt: 443
Melting Pt: 278 °C
MDL Number: MFCD00004142
CAS Number: 198-55-0
Merck Index: 12,07325
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac9b8d3c5af9d9807c1ced6d476d2cd598e5c834&vid=3749053ee2278bf0ba2dc838805f8b8a80b6a0cb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR