1-Pyreneboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
P1625-1G
P1625-5G
P1625-25G
TCP1625-1GEA
39
USD
TCP1625-1G
TCP1625-5G
TCP1625-25G
1-Pyreneboronic acid (contains varying amounts of Anhydride)
1-Pyreneboronic acid
CAS Number: 164461-18-1
Molecular Formula: C16H11BO2
Molecular Weight: 246.07
Form: Crystal
Color: White
Melting point (°C): 251
Formula:
C₁₆H₁₁BO₂ Melting Pt: 251 °C |
MDL Number:
MFCD04974062 CAS Number: 164461-18-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...