trans-4-(4-Propylcyclohexyl)benzonitrile ≥98.0%
Supplier: TCI America
Synonyms:
4-(trans-4-propylcyclohexyl)benzonitrile
P1617-5G
TCP1617-5GEA
132
USD
TCP1617-5G
trans-4-(4-Propylcyclohexyl)benzonitrile ≥98.0%
trans-4-(4-Propylcyclohexyl)benzonitrile
CAS Number: 61203-99-4
MDL Number: MFCD04038799
Molecular Formula: C16H21N
Molecular Weight: 227.35
Purity/Analysis Method:
Product Categories: Materials Chemistry, Liquid Crystal Materials (Optically Functional Materials)
Other Product Names / Synonyms: 1-Cyano-4-(trans-4-propylcyclohexyl)benzene
Formula:
C₁₆H₂₁N MW: 227.3446 g/mol Melting Pt: 45 °C |
MDL Number:
MFCD04038799 CAS Number: 61203-99-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...