Potassium permanganate in solution (contains K2CO3, NaOH)

Supplier: TCI America

Synonyms: Permanganic acid potassium salt

P1483-500ML
TCP1483-500MLEA 137.86 USD
TCP1483-500ML
Potassium permanganate in solution (contains K2CO3, NaOH)
Potassium permanganate

[for TLC Stain]
CAS Number: 7722-64-7
MDL Number: MFCD00011364
Molecular Formula: KMnO4
Molecular Weight: 158.03
Form: Liquid
Color: Deep Purple

Formula: KMnO₄
MW: 158.03 g/mol
MDL Number: MFCD00011364
CAS Number: 7722-64-7
UN: 1490
ADR: 5.1 II
Merck Index: 12,07824

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR