(R,R)-(-)-2,4-Pentanediol ≥98.0%
Supplier: TCI America
Synonyms:
(R,R)-(-)-2,4-Dihydroxy pentane, (2R,4R)-(-)-2,4-Pentanediol, (2R,4R)-Pentane-2,4-diol
P1411-1G
TCP1411-1GEA
155
USD
TCP1411-1G
(R,R)-(-)-2,4-Pentanediol ≥98.0%
(R,R)-(-)-2,4-Pentanediol
CAS Number: 42075-32-1
MDL Number: MFCD00063893
Molecular Formula: C5H12O2
Molecular Weight: 104.15
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (2R,4R)-(-)-2,4-Dihydroxypentane
Formula:
C₅H₁₂O₂ Melting Pt: 50 °C Flash Pt: 101 °C |
MDL Number:
MFCD00063893 CAS Number: 42075-32-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...