o,o,o-Tritolyl phosphate ≥97.0%

Supplier: TCI America

P1331-25G P1331-5G
TCP1331-025GEA 664.9 USD
TCP1331-025G TCP1331-5G
o,o,o-Tritolyl phosphate ≥97.0%
o,o,o-Tritolyl phosphate

CAS Number: 78-30-8 MDL Number: MFCD00041906 Molecular Formula: C21H21O4P Molecular Weight: 368.37 Purity/Analysis Method: 97.0% (GC) Form: Clear Liquid Color: Slightly Pale Yellow Boiling point (°C): 410 Flash Point (°C): 225 Specific Gravity (20/20): 1.18


Product Categories: Materials Chemistry, Secondary Battery Research Reagents, Resin Additives, Resin Additives Other Product Names / Synonyms: Phosphoric Acid Tri-o-tolyl Ester, Phosphoric Acid Tri-o-cresyl Ester, Tri-o-tolyl Phosphate

Formula: C₂₁H₂₁O₄P
Boiling Pt: 410 °C
Melting Pt: 11 °C
Density: 1.18 (20 °C)
Flash Pt: 225 °C
MDL Number: MFCD00041906
CAS Number: 78-30-8
EINECS: 201-103-5
UN: 2574
ADR: 6.1,II
Merck Index: 12,09893

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR