(S)-(+)-β-Aminophenethyl alcohol ≥97.0%
Supplier: TCI America
Synonyms:
(S)-(+)-2-Phenylglycinol, L-Phenylglycinol, (S)-2-Amino-2-phenylethanol
P1294-25G
P1294-5G
TCP1294-25GEA
67.13
USD
TCP1294-25G
TCP1294-005G
(S)-(+)-β-Aminophenethyl alcohol ≥97.0%
(S)-(+)-β-Aminophenethyl alcohol
CAS Number: 20989-17-7
MDL Number: MFCD00064404
Molecular Formula: C8H11NO
Molecular Weight: 137.18
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-(+)-2-Amino-2-phenylethanol
Formula:
C₈H₁₁NO Melting Pt: 74 °C |
MDL Number:
MFCD00064404 CAS Number: 20989-17-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...