1,1,1,2,2,3,3-Heptafluoro-3-[(trifluorovinyl)oxy]propane ≥98.0% for synthesis

Supplier: TCI America

Synonyms: Perfluoropropoxyethylene, (Heptafluoropropoxy)trifluoroethene, Perfluoro(propyl vnyl ether)

P1224-10G
TCP1224-010GEA 50 USD
TCP1224-010G
1,1,1,2,2,3,3-Heptafluoro-3-[(trifluorovinyl)oxy]propane ≥98.0% for synthesis
1,1,1,2,2,3,3-Heptafluoro-3-[(trifluorovinyl)oxy]propane

CAS Number: 1623-05-8 MDL Number: MFCD00191577 Molecular Formula: C5F10O Molecular Weight: 266.04 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Color: Colorless Boiling point (°C): 36 Specific Gravity (20/20): 1.56 Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorous Compounds Other Product Names / Synonyms: Perfluoro(propyl Vinyl Ether)

Formula: C₅F₁₀O
Boiling Pt: 36 °C
Density: 1.56 (20 °C)
MDL Number: MFCD00191577
CAS Number: 1623-05-8
EINECS: 216-600-2

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR