Tetrabenzyl pyrophosphate ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: Tetrabenzyl diphosphate

P1223-1G
TCP1223-1GEA 73.2 USD
TCP1223-1G
Tetrabenzyl pyrophosphate ≥98.0% (by HPLC)
Tetrabenzyl pyrophosphate

CAS Number: 990-91-0 MDL Number: MFCD00051941 Molecular Formula: C28H28O7P2 Molecular Weight: 538.47 Purity/Analysis Method: 98.0% (HPLC) Form: Crystal Color: White Melting point (°C): 64 Storage Temperature: 0°C


Product Categories: Synthetic Organic Chemistry, Phosphorylation, Nucleic Acid Synthesis Other Product Names / Synonyms: Pyrophosphoric Acid Tetrabenzyl Ester, Tetrabenzyl Diphosphate

Formula: C₂₈H₂₈O₇P₂
MW: 538.48 g/mol
Melting Pt: 64 °C
MDL Number: MFCD00051941
CAS Number: 990-91-0
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=93abdcf02ba91ecfe726df85f4ccc917d6472cf9&vid=93abdcf02ba91ecfe726df85f4ccc917d6472cf9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR