(R)-(-)-Hydratropic acid ≥98.0%

Supplier: TCI America
Warning

Synonyms: (R)-(-)-2-Phenylpropanoic acid, (R)-(-)-2-Phenylpropionic acid, (R)-2-Phenylpropanoic acid

P1219-5G P1219-1G
TCP1219-5GEA 328 USD
TCP1219-5G TCP1219-001G
(R)-(-)-Hydratropic acid ≥98.0%
(R)-(-)-Hydratropic acid

CAS Number: 7782-26-5 MDL Number: MFCD00063140 Molecular Formula: C9H10O2 Molecular Weight: 150.18 Purity/Analysis Method: 98.0% (GC,T) Form: Crystal Color: White Flash Point (°C): 110 Freezing point (°C): 29 Specific rotation [a]20/D: -80 deg (C=3, EtOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Analytical Science, e.e. / Absolute Configuration Determination (NMR) Other Product Names / Synonyms: (R)-(-)-Hydratropic Acid

Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 257
Melting Pt: 29…32 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063140
CAS Number: 7782-26-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=760b2dd8d41e2a4f2e97d25f2b6a61d9aca86f89&vid=760b2dd8d41e2a4f2e97d25f2b6a61d9aca86f89&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR