N-(1-Pyrenyl)maleimide ≥97.0%
Supplier: TCI America
Synonyms:
1-(Pyren-1-yl)-1H-pyrrole-2,5-dione
P1214-250MG
TCP1214-250MGEA
126
USD
TCP1214-250MG
N-(1-Pyrenyl)maleimide ≥97.0%
N-(1-Pyrenyl)maleimide
CAS Number: 42189-56-0
MDL Number: MFCD00049301
Molecular Formula: C20H11NO2
Molecular Weight: 297.31
Purity/Analysis Method:
Formula:
C₂₀H₁₁NO₂ MW: 297.31 g/mol |
MDL Number:
MFCD00049301 CAS Number: 42189-56-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...