9,10-Bis(phenylethynyl)anthracene ≥98.0% (by HPLC)
Supplier: TCI America
P1137-1G
P1137-5G
TCP1137-001GEA
58
USD
TCP1137-001G
TCP1137-5G
9,10-Bis(phenylethynyl)anthracene ≥98.0% (by HPLC)
9,10-Bis(phenylethynyl)anthracene
CAS Number: 10075-85-1
MDL Number: MFCD00012050
Molecular Formula: C30H18
Molecular Weight: 378.47
Purity/Analysis Method:
Formula:
C₃₀H₁₈ Melting Pt: 253 °C |
MDL Number:
MFCD00012050 CAS Number: 10075-85-1 EINECS: 233-210-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...