Tris(1H,1H,5H-octafluoropentyl)phosphate ≥85.0%
Supplier: TCI America
CAS Number: 355-86-2
MDL Number: MFCD00054654
Molecular Formula: C15H9F24O4P
Molecular Weight: 740.17
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: Phosphoric Acid Tris(1H,1H,5H-octafluoropentyl) Ester
Formula:
C₁₅H₉F₂₄O₄P Boiling Pt: 154 °C Density: 1.77 (20 °C) Flash Pt: 100 °C |
MDL Number:
MFCD00054654 CAS Number: 355-86-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...