Pentafluoropropionic acid ≥98.0%
Supplier: TCI America
Synonyms:
Perfluoropropionic acid, Pentafluoropropanoic acid, Perfluoropropanoic acid, 2,2,3,3,3-Pentafluoropropionic acid
P1125-100G
P1125-25G
TCP1125-100GEA
404.98
USD
TCP1125-100G
TCP1125-25G
Pentafluoropropionic acid ≥98.0%
Pentafluoropropionic acid
CAS Number: 422-64-0
MDL Number: MFCD00004170
Molecular Formula: C3HF5O2
Molecular Weight: 164.03
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks
Other Product Names / Synonyms: Perfluoropropionic Acid
Formula:
C₃HF₅O₂ Boiling Pt: 98 °C Density: 1.58 (20 °C) |
MDL Number:
MFCD00004170 CAS Number: 422-64-0 UN: 3265 ADR: 8,II |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...