Pyrene ≥97.0%

Supplier: TCI America

P1104-25G P1104-500G P1104-100G
TCP1104-25GEA 62.07 USD
TCP1104-25G TCP1104-500G TCP1104-100G
Pyrene ≥97.0%
Pyrene

CAS Number: 129-00-0 MDL Number: MFCD00004136 Molecular Formula: C16H10 Molecular Weight: 202.26 Purity/Analysis Method: 97.0% (GC) Form: Crystal Color: White Boiling point (°C): 404 Melting point (°C): 150 Flash Point (°C): 199 Lambda max.: 330 nm (EtOH)


Product Categories: Materials Chemistry, Nanocarbon Unit Structures (Nanocarbon Materials) Other Product Names / Synonyms: Benzo[def]phenanthrene

Formula: C₁₆H₁₀
Boiling Pt: 404 °C
Melting Pt: 150 °C
Flash Pt: 199 °C
MDL Number: MFCD00004136
CAS Number: 129-00-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87918eb4de97c8725e2efe6b826198a273e81dfe&vid=87918eb4de97c8725e2efe6b826198a273e81dfe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR